Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, 1 micron, dry form

$415.86 - $415.86

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
Name Note Dry form
Molecular Formula [-CH2CH(C6H5)-]n
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43711ME

  SDS
alfa aesar™
43711ME
500mg Each for $415.86
Description & Specifications

Specifications

Physical Form Microsphere
TSCA TSCA
Name Note Dry form
CAS 9003-53-6
Chemical Name or Material Polystyrene latex microsphere
Particle Size 1 Micron
MDL Number MFCD00084450
Odor Odorless
Quantity 500mg
Molecular Formula [-CH2CH(C6H5)-]n