Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, 1 micron, 10 wt% dispersion in water

$781.96 - $781.96

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA419177C

  SDS
alfa aesar™
419177C
15mL Each for $781.96
Description & Specifications

Specifications

Color White
Chemical Name or Material Polystyrene latex microsphere
TSCA TSCA
Physical Form Dispersion
MDL Number MFCD00084450
CAS 9003-53-6
Odor Odorless
Particle Size 1 Micron
Concentration 10 wt% dispersion in water
Quantity 15mL