Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, 0.75 micron, 2.5 wt% dispersion in water

$263.69 - $263.69

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
Molecular Formula [-CH2CH(C6H5)-]n
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA427424Y

  SDS
alfa aesar™
427424Y
15mL Each for $263.69
Description & Specifications

Specifications

Particle Size 0.75 Micron
Odor Odorless
RTECS Number WL6475000
Concentration 2.5 wt% dispersion in water
Recommended Storage 2°C to 8°C
Physical Form Microsphere
Molecular Formula [-CH2CH(C6H5)-]n
MDL Number MFCD00084450
Color Opalescent
Quantity 15mL
Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
TSCA TSCA
Safety and Handling

2°C to 8°C