Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, 0.50 micron, 2.5 wt% dispersion in water

$253.14 - $253.14

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA427144Y

  SDS
alfa aesar™
427144Y
15mL Each for $253.14
Description & Specifications

Specifications

Color Opalescent
TSCA TSCA
Physical Form Dispersion
Odor Odorless
Storage Note 1 2°C to 8°C
Quantity 15mL
CAS 9003-53-6
Particle Size 0.50 Micron
Chemical Name or Material Polystyrene latex microsphere
MDL Number MFCD00084450
Concentration 2.5 wt% dispersion in water
RTECS Number WL6475000
Safety and Handling

2°C to 8°C