Please login to your online account to display your discounted pricing

Alfa Aesar™ Polystyrene latex microsphere, 0.20 micron, 2.5 wt% dispersion in water

$229.03 - $229.03

Chemical Identifiers

Chemical Name or Material Polystyrene latex microsphere
CAS 9003-53-6
MDL Number MFCD00084450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA427134Y

  SDS
alfa aesar™
427134Y
15mL Each for $229.03
Description & Specifications

Specifications

Chemical Name or Material Polystyrene latex microsphere
Storage Note 1 2°C to 8°C
Odor Odorless
Particle Size 0.20 Micron
Concentration 2.5 wt% dispersion in water
TSCA TSCA
Color White
RTECS Number WL6475000
MDL Number MFCD00084450
CAS 9003-53-6
Physical Form Dispersion
Quantity 15mL
Safety and Handling

2°C to 8°C