missing translation for 'onlineSavingsMsg'
Learn More

Polystyrene, Analytical Standard, For GPC, 2,000,000, MilliporeSigma™ Supelco™

Catalog No. 111004580
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004580 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004580 Supplier Merck Emd Millipore Supplier No. 3278161G
Only null left

Polystyrene (for GPC, 2,000,000), a synthetic thermoplastic, is used widely for various applications owing to its properties such as cheap cost, rigidity, low specific weight, high chemical resistance, mechanical flexibility, biocompatibility and good processability.

Specifications

Packaging Glass Bottle
Physical Form Fibers
Linear Formula [CH2CH(C6H5)]n
MDL Number MFCD00084450
Synonym PS
Quantity 1 g
Grade GPC