Please login to your online account to display your discounted pricing

Polystyrene: 2% Divinylbenzene Copolymer Beads 200-400 mesh, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: fisher scientific acros  418105000

 View more versions of this product

Catalog No. AC418105000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Beads
Quantity 500g
Color White to off-white
TSCA TSCA
Chemical Name or Material Polystyrene: 2% divinylbenzene copolymer beads
Name Note 200-400 mesh
MDL Number MFCD00146442
CAS 9003-70-7
Infrared Spectrum Authentic
Loss on Drying 2% max. (105°C)
Mesh Size 200 to 400 mesh
Packaging Glass bottle

Specifications

Chemical Name or Material Polystyrene: 2% divinylbenzene copolymer beads
Name Note 200-400 mesh
CAS 9003-70-7
MDL Number MFCD00146442