Please login to your online account to display your discounted pricing

Polystyrene: 2% Divinylbenzene Copolymer Beads 200-400 mesh, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Polystyrene: 2% divinylbenzene copolymer beads
Name Note 200-400 mesh
CAS 9003-70-7
MDL Number MFCD00146442
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC418105000

  SDS
fisher scientific acros
418105000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Beads
Quantity 500g
Color White to off-white
TSCA TSCA
Chemical Name or Material Polystyrene: 2% divinylbenzene copolymer beads
Name Note 200-400 mesh
MDL Number MFCD00146442
CAS 9003-70-7
Infrared Spectrum Authentic
Loss on Drying 2% max. (105°C)
Mesh Size 200 to 400 mesh
Packaging Glass bottle