Please login to your online account to display your discounted pricing

Polysorbate 80, Vegetable, Multi-Compendial, N.F., J.T.Baker™


Quantity 200mL
Packaging Narrow Mouth Amber Glass Bottle

Manufacturer: avantor performance  450004

 View more versions of this product

Catalog No. 02003655

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Multi-Compendial/NF
CAS 9005-65-6
Chemical Name or Material Polysorbate 80, Vegetable
Packaging Narrow Mouth Amber Glass Bottle
Flash Point 148.00°C
Quantity 200mL
Specific Gravity 1L = 1.08kg

Specifications

Chemical Name or Material Polysorbate 80, Vegetable
CAS 9005-65-6
Purity Grade Multi-Compendial/NF