missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polysorbate 80, Vegetable, Multi-Compendial, N.F., J.T. Baker™
Supplier: Avantor J.T.Baker 450006
Specifications
Polysorbate 80, Vegetable | |
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | |
1 L | |
HDTIFOGXOGLRCB-KTKRTIGZNA-N | |
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate | |
Multi-Compendial/NF | |
1L = 1.08kg |
9005-65-6 | |
MFCD00082107 | |
148.00°C | |
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
604.82 | |
Narrow Mouth Amber Glass Bottle |
Chemical Identifiers
9005-65-6 | |
604.82 | |
HDTIFOGXOGLRCB-KTKRTIGZNA-N | |
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | |
MFCD00082107 | |
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate |