Please login to your online account to display your discounted pricing

Polysorbate 80, Multi-Compendial, N.F., J.T.Baker™


Quantity 4L
Packaging Narrow Mouth Amber Glass Bottle

Manufacturer: avantor performance  411703

 View more versions of this product

Catalog No. 02003662

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Multi-Compendial/NF
Chemical Name or Material Polysorbate 80
CAS 9005-65-6
Quantity 4L
Packaging Narrow Mouth Amber Glass Bottle
Flash Point 148.00°C
Specific Gravity 1L = 1.08kg

Specifications

Chemical Name or Material Polysorbate 80
CAS 9005-65-6
Purity Grade Multi-Compendial/NF