Please login to your online account to display your discounted pricing

Alfa Aesar™ Polysorbate 80


Quantity 500mL

Manufacturer: alfa aesar™  L13315AP

 View more versions of this product

Catalog No. AAL13315AP

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Polysorbate 80
TSCA TSCA
MDL Number MFCD00082107
Merck Index 14,7582
Boiling Point >100°C
Flash Point >149°C (300°F)
CAS 9005-65-6
Synonym Polyoxyethylene sorbitan monooleate
Quantity 500mL
RTECS Number WG2932500

Specifications

Chemical Name or Material Polysorbate 80
CAS 9005-65-6
MDL Number MFCD00082107
Merck Index 14,7582
Synonym Polyoxyethylene sorbitan monooleate