Learn More
Polysorbate 60, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L15031AP
Description
Polysorbate 60 is used in food, medicine, plastic, cosmetics and other industries. It is an excellent emulgator and one of elements to constitute polyacrylonitrile fiber spinning oil. It also used as softening agent after processing fiber and eliminate fiber static and improve its softness.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Polysorbate 60 | |
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6 | |
1.08 | |
Polyoxyethylene sorbitan monostearate | |
500 mL | |
CCCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
522.68 |
9005-67-8 | |
MFCD01780110 | |
14,7582 | |
Soluble in water and alcohols. Insoluble in oils. | |
DEKFZWMENCCOFU-UHFFFAOYNA-N | |
2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate | |
>150°C (302°F) |
Chemical Identifiers
9005-67-8 | |
522.68 | |
DEKFZWMENCCOFU-UHFFFAOYNA-N | |
2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6 | |
MFCD01780110 | |
Polyoxyethylene sorbitan monostearate | |
CCCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
Safety and Handling
EINECSNumber : 000-000-0
RTECSNumber : WG2934000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only