Please login to your online account to display your discounted pricing

Polysorbate 20, Multi-Compendial, N.F., J.T.Baker™


Quantity 1L
Packaging Narrow Mouth Amber Glass Bottle

Manufacturer: avantor performance  411606

 View more versions of this product

Catalog No. 02003665

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Multi-Compendial/NF
Chemical Name or Material Polysorbate 20
Flash Point 148.00°C
Quantity 1L
Packaging Narrow Mouth Amber Glass Bottle
CAS 9005-64-5
Specific Gravity 1L = 1.11kg

Specifications

Chemical Name or Material Polysorbate 20
CAS 9005-64-5
Purity Grade Multi-Compendial/NF