missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polysorbate 20, MP Biomedicals™
Manufacturer: Mp Biomedicals 0219484150
Specifications
Polysorbate 20 | |
1.095 g/mL | |
C26H50O10 | |
50mL | |
HMFKFHLTUCJZJO-UHFFFAOYNA-N | |
2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate | |
443314 |
9005-64-5 | |
Yellow | |
MFCD00165986 | |
Polyoxyethylenesorbitan monolaurate, Tween 20 | |
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
522.68 | |
Oily Liquid |
Chemical Identifiers
9005-64-5 | |
522.68 | |
HMFKFHLTUCJZJO-UHFFFAOYNA-N | |
443314 | |
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
C26H50O10 | |
MFCD00165986 | |
Polyoxyethylenesorbitan monolaurate, Tween 20 | |
2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |