missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polysorbate 20, Electrophoresis Reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J66278AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Polysorbate 20 | |
Electrophoresis Reagent | |
Liquid | |
1.105 g/mL | |
MFCD00165986 | |
Polyoxyethylene sorbitan monolaurate; Tween™ 20 | |
HMFKFHLTUCJZJO-UHFFFAOYNA-N | |
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate | |
443314 |
Colorless to Yellow | |
9005-64-5 | |
>100°C | |
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | |
100 mL | |
>110°C (230°F) | |
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
522.68 | |
1.47 |
Chemical Identifiers
9005-64-5 | |
522.68 | |
HMFKFHLTUCJZJO-UHFFFAOYNA-N | |
443314 | |
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | |
MFCD00165986 | |
Polyoxyethylene sorbitan monolaurate; Tween™ 20 | |
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate |
Safety and Handling
EINECSNumber : 500-018-3
RTECSNumber : TR7400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only