Please login to your online account to display your discounted pricing

Alfa Aesar™ Polysorbate 20


Quantity 100mL

Manufacturer: alfa aesar™  L15029AE

 View more versions of this product

Catalog No. AAL15029AE

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Polysorbate 20
CAS 9005-64-5
TSCA TSCA
Molecular Formula C58H114O26
Synonym Polyoxyethylene sorbitan monolaurate
Boiling Point >100°C
Quantity 100mL
Flash Point >110°C (230°F)
RTECS Number TR7400000
MDL Number MFCD00165986

Specifications

Chemical Name or Material Polysorbate 20
CAS 9005-64-5
Molecular Formula C58H114O26
MDL Number MFCD00165986
Synonym Polyoxyethylene sorbitan monolaurate