missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polyseed NX™, For BOD, Reagent Grade, LabChem™

$396.38 - $396.38

Chemical Identifiers

Chemical Name or Material Polyseed NX™, For BOD
Grade Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC1889050 Lab Chem
LC1889050
50 Capsules Each for $396.38
Specifications Description & Specifications

Chemical Identifiers

Polyseed NX™, For BOD
Reagent Grade

Specifications

Liquid
Bottle
Room Temperature
50 Capsules
Reagent Grade
Passes Test
Soluble in water
Contains hexavalent Chromium
Polyseed NX™, For BOD

Safety and Handling

GHS H Statement
Product is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.