missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polyseed™, For BOD, Reagent Grade, LabChem™

$314.74 - $314.74

Chemical Identifiers

Chemical Name or Material Polyseed™, For BOD
Grade Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC1888050 Lab Chem
LC1888050
50 Capsules Each for $314.74
Specifications Description & Specifications

Chemical Identifiers

Polyseed™, For BOD
Reagent Grade

Specifications

Soluble in water
50 Capsules
Passes Test
Room Temperature
Reagent Grade
Liquid
Bottle
Contains hexavalent Chromium
Polyseed™, For BOD

Safety and Handling

GHS H Statement
Product is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.