Please login to your online account to display your discounted pricing

Polyseed™ BOD and Polyseed-NX™ CBOD Capsules


Produce fast, economical BOD analyses with consistent results

Manufacturer: interlab  P110

 View more versions of this product

Catalog No. 13-297-200

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Certifications/Compliance EPA
For Use With (Application) Ideal for use in a broad range of areas involving degradation of industrial and municipal waste
Includes Specialized microbial cultures
Shelf Life 2 Years
Yield Up to 200 tests
Product Type Polyseed BOD Capsules
Accessories

2 Years