missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Polypropylene Storage Vial with Screw Cap

Eisco™ Polypropylene Storage Vial with Screw Cap provides leak free storage of liquids, beads, grains or powders and allows visibility of the contents for display or ease of use.

$319.94 - $319.94

Specifications

Color Clear
Material Polypropylene
Sterility Non-sterile
For Use With (Application) Storage
Closure Material polypropylene
View More Specs

Products 3
Catalog Number Mfr. No. Volume (Metric) Height (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Height (Metric) Price Quantity  
S29358
View Documents Promotion Details
Eisco™
BI0390A
1 mL 10.2 cm
Each for $319.94
 
S29359
View Documents Promotion Details
Eisco™
BI0390B
2 mL 10 cm
Each for $319.94
 
S29360
View Documents Promotion Details
Eisco™
BI0390C
5 mL 7.6 cm
Each for $319.94
 
Description

Description

  • Rugged and durable polypropylene storage vials
  • Clear for easy viewing of contents
  • Leak proof screw top
  • Pack of 500

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

Clear
Non-sterile
polypropylene
Screw Cap
Polypropylene
Storage
Autoclavable
500
SDS
missing translation for 'documents'

missing translation for 'documents'