Please login to your online account to display your discounted pricing

Alfa Aesar™ Polypropylene sheet, 6.36mm (0.25 in.) thick

$42.34 - $42.34

Chemical Identifiers

Chemical Name or Material Polypropylene sheet
CAS 9003-07-0
Name Note 6.36mm (0.25 in.) thick
MDL Number MFCD00084447
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45174HB

  SDS
alfa aesar™
45174HB
300x300mm Each for $42.34
Description & Specifications

Specifications

MDL Number MFCD00084447
Quantity 300x300mm
TSCA TSCA
CAS 9003-07-0
RTECS Number UD1842000
Chemical Name or Material Polypropylene sheet
Name Note 6.36mm (0.25 in.) thick