missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polypropylene sheet, 3.18mm (0.125mm) thick

Manufacturer:  Alfa Aesar™ 45196HB

Catalog No. AA45196HB



Specifications

Specifications

Polypropylene sheet
9003-07-0
MFCD00084447
NSYDMBURIUSUDH-RTWAWAEBSA-N
12-[(2S,3R)-3-octyloxiran-2-yl]dodecanoic acid
76958
3.18mm (0.125mm) thick
C22H42O3
12-2s,3r-3-octyloxiran-2-yl dodecanoic acid, 12-3-octyl-2-oxiranyl dodecanoic acid #, 2-oxiranedodecanoic acid, 3-octyl-, 2r,3s-rel, 3beta-octyloxirane-2beta-dodecanoic acid, oxiranedodecanoic acid, 3-octyl-, 2r,3s-rel, oxiranedodecanoic acid, 3-octyl-, cis, polypropylene, special for nanometer polypropylene pipe material
CCCCCCCCC1C(O1)CCCCCCCCCCCC(=O)O
354.575
300 x 300mm

Chemical Identifiers

9003-07-0
354.575
NSYDMBURIUSUDH-RTWAWAEBSA-N
76958
CCCCCCCCC1C(O1)CCCCCCCCCCCC(=O)O
C22H42O3
MFCD00084447
12-2s,3r-3-octyloxiran-2-yl dodecanoic acid, 12-3-octyl-2-oxiranyl dodecanoic acid #, 2-oxiranedodecanoic acid, 3-octyl-, 2r,3s-rel, 3beta-octyloxirane-2beta-dodecanoic acid, oxiranedodecanoic acid, 3-octyl-, 2r,3s-rel, oxiranedodecanoic acid, 3-octyl-, cis, polypropylene, special for nanometer polypropylene pipe material
12-[(2S,3R)-3-octyloxiran-2-yl]dodecanoic acid
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents