Please login to your online account to display your discounted pricing

Alfa Aesar™ Polypropylene sheet, 3.18mm (0.125mm) thick

$38.18 - $38.18

Chemical Identifiers

Chemical Name or Material Polypropylene sheet
Name Note 3.18mm (0.125mm) thick
MDL Number MFCD00084447
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45196HB

 
alfa aesar™
45196HB
300x300mm Each for $38.18
Description & Specifications

Specifications

Chemical Name or Material Polypropylene sheet
TSCA TSCA
Name Note 3.18mm (0.125mm) thick
MDL Number MFCD00084447
Quantity 300x300mm