Please login to your online account to display your discounted pricing

Alfa Aesar™ Polypropylene sheet, 12.7mm (0.5 in.) thick

$59.38 - $59.38

Chemical Identifiers

Chemical Name or Material Polypropylene sheet
Name Note 12.7mm (0.5 in.) thick
MDL Number MFCD00084447
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45215HB

  SDS
alfa aesar™
45215HB
300x300mm Each for $59.38
Description & Specifications

Specifications

Chemical Name or Material Polypropylene sheet
TSCA TSCA
Name Note 12.7mm (0.5 in.) thick
MDL Number MFCD00084447
Quantity 300x300mm