missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Keystone Safety Polypropylene (Light Vinyl) Shoe Covers - Non-Skid

$104.43 - $104.43

Specifications

Material Polypropylene
Coating Vinyl
Color Blue
Sole Type Non-skid
Weight (English) 5 lb.
View More Specs

Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19055181
View Documents Promotion Details
Keystone Safety
SCCPEXL
X-Large
Case of 300 Each for $104.43
 
Description

Description

Specifications

Specifications

Polypropylene
Blue
5 lb.
Fluid
Vinyl
Non-skid
2.26 kg
Controlled Environments Shoe Covers
SDS
missing translation for 'documents'

missing translation for 'documents'