missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polypropylene glycol 2000, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L14699AE

 View more versions of this product

Catalog No. AAL14699AE



Description

Description

Used to make polyester compounds, absorb extra water and maintain moisture in certain medicines, cosmetics, or food products and as a base for deicing solutions. Propylene glycol is used by the chemical, food, and pharmaceutical industries as an antifreeze. It is a solvent for food colors and flavors, and in the paint and plastics industries. Propylene glycol is also used to create artificial smoke or fog used in fire-fighting training and in theatrical productions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Polypropylene glycol 2000
(C3H6O)n
1.002
Miscible with water,ethanol,diethyl ether and chloroform.
DNIAPMSPPWPWGF-UHFFFAOYNA-N
2-(2-hydroxypropoxy)propan-1-ol
32881
194°C (381°F)
25322-69-4
MFCD00084446
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
100 mL
[H]OC(C)CO
134.18
2000
1.451

Chemical Identifiers

25322-69-4
134.18
DNIAPMSPPWPWGF-UHFFFAOYNA-N
32881
[H]OC(C)CO
(C3H6O)n
MFCD00084446
polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
2-(2-hydroxypropoxy)propan-1-ol
Safety and Handling

Safety and Handling

EINECSNumber : 500-039-8

RTECSNumber : TR6130000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only