Dynalon™ Polypropylene Flasks
Your best choice for basic chemistry labs.
$20.46 - $30.21
Specifications
Description | Style: Flask, Erlenmeyer |
---|---|
Material | Polypropylene |
Autoclavable | Autoclavable |
Description
- Autoclavable
- Sturdy and chemically resistant
- With molded-in graduations
Specifications
Style: Flask, Erlenmeyer | |
Polypropylene | |
Autoclavable |