missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Polypropylene Filtering Flasks
Buchner filtering flask made of high quality, chemical resistant polypropylene with a conical shape to prevent spills
$76.18 - $95.44
Specifications
Material | Polypropylene |
---|---|
Neck Style | Beaded Rim |
Product Type | Filtering Flask |
Shape | Conical |
Autoclavable | Autoclavable |
Description
- Made of high quality, chemical resistant polypropylene
- Conical shape, beaded rim to prevent drips
- Autoclavable at 121°C
- Microwavable
- Ungraduated
Specifications
Polypropylene | |
Filtering Flask | |
Autoclavable | |
No | |
Non-sterile |
Beaded Rim | |
Conical | |
Reusable | |
Narrow | |
Flat |