missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polyphosphoric acid, pure, >84% phosphate (as P2O5), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 196950025

 View more versions of this product

Catalog No. AC196950025




Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Polyphosphoric acid
550.0°C
Plastic bottle
2.5 kg
Viscous Liquid
Solubility in water: may decompose. Other solubilities: insoluble in hydrocarbons and halogenated hydrocar, soluble in alcohols
[H]OP(O)(O)=O
≥82% (as P2O5) (Alkalimetry)
8017-16-1
2.0500g/mL
2.05
(HO3P)n
MFCD00084480
NBIIXXVUZAFLBC-UHFFFAOYSA-N
Polyphosphoric acid
Pure

Chemical Identifiers

8017-16-1
MFCD00084480
Polyphosphoric acid
(HO3P)n
NBIIXXVUZAFLBC-UHFFFAOYSA-N
[H]OP(O)(O)=O
Safety and Handling

Safety and Handling

Recommended Storage : May darken over time

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only