missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyphenylmethylsiloxane, MW 2500-2700, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04456918
Description
A biochemical for proteomics research. A novel coating for solid-phase microextraction. A polyphenylmethylsiloxane for use in skincare, suncare, color cosmetic, hair care, and antiperspirant / deodorant applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Polyphenylmethylsiloxane | |
Liquid | |
C7H8OSi | |
methylphenylsilanone, polyphenylmethylsiloxane, methylphenyl oxo silane, silane, methyloxophenyl, methyl phenyl silanone | |
50 g | |
C[Si](=O)C1=CC=CC=C1 | |
136.23 | |
300°C (572°F) |
M.W. 2,500 to 2,700 | |
9005-12-3 | |
MFCD00146731 | |
Insoluble in water. | |
IXKNJYSAWNJQQY-UHFFFAOYSA-N | |
methyl-oxo-phenylsilane | |
3032548 |
Chemical Identifiers
9005-12-3 | |
136.23 | |
MFCD00146731 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only