missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyphenylmethylsiloxane, MW 2500-2700

Manufacturer:  Alfa Aesar™ 4456930

 View more versions of this product

Catalog No. AA4456930



Specifications

Specifications

Polyphenylmethylsiloxane
9005-12-3
MFCD00146731
Insoluble in water.
C[Si](=O)C1=CC=CC=C1
136.23
Liquid
300°C (572°F)
M.W. 2,500 to 2,700
C7H8OSi
methyl phenyl silanone
IXKNJYSAWNJQQY-UHFFFAOYSA-N
methyl-oxo-phenylsilane
3032548
250g

Chemical Identifiers

9005-12-3
136.23
IXKNJYSAWNJQQY-UHFFFAOYSA-N
3032548
C[Si](=O)C1=CC=CC=C1
C7H8OSi
MFCD00146731
methyl phenyl silanone
methyl-oxo-phenylsilane
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents