missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyoxyethylene bis(amine), M.W. 8,000, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 046903MD
Description
It is used as PEG modifier, its main products include high-qualified intermediates, various of amino acid and APIs. This Polymer used in the preparation of enzyme conjugates soluble in organic solvents. Promising drug carrier.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
White | |
M.W. 8,000 | |
24991-53-5 | |
MFCD00132004 | |
It is soluble in organic solvents. It is insoluble in water. | |
Air Sensitive |
Polyoxyethylene bis (amine) | |
Granular Solid | |
H2NCH2CH2(OCH2CH2)nNH2 | |
PEG-diamine | |
250 mg |
Chemical Identifiers
24991-53-5 | |
H2NCH2CH2(OCH2CH2)nNH2 | |
MFCD00132004 | |
PEG-diamine |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only