missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyoxyethylene bis(amine), M.W. 3,400, Thermo Scientific Chemicals
$75.55 - $971.52
Chemical Identifiers
CAS | 24991-53-5 |
---|---|
Molecular Formula | H2NCH2CH2(OCH2CH2)nNH2 |
MDL Number | MFCD00132004 |
Synonym | PEG-diamine |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA46973MD
|
Thermo Scientific Chemicals
046973MD |
250 mg |
Each for $75.55
|
|
|||||
AA4697303
|
Thermo Scientific Chemicals
04697303 |
1 g |
Each for $225.03
|
|
|||||
AA4697306
|
Thermo Scientific Chemicals
04697306 |
5 g |
Each for $971.52
|
|
|||||
Description
Polyoxyethylene bis(amine) with HCl, it act as reduction agent; stereo selective allylation of carbonyl compounds; in situ generation of tinenolates for directed aldol reactions. It is used in the preparation of enzyme conjugates, promising drug carrier.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
24991-53-5 | |
H2NCH2CH2(OCH2CH2)nNH2 | |
MFCD00132004 | |
PEG-diamine |
Specifications
White | |
24991-53-5 | |
MFCD00132004 | |
Soluble in organic solvents. | |
Air Sensitive |
Granular Solid | |
H2NCH2CH2(OCH2CH2)nNH2 | |
PEG-diamine | |
250 mg | |
Polyoxyethylene bis (amine), M.W. 3,400 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only