Alfa Aesar™ Polyoxyethylene bis(amine), M.W. 1,000
Manufacturer:  Alfa Aesar™ 46833MD
 View more versions of this product
Catalog No. AA46833MD
Specifications
Specifications
Polyoxyethylene bis (amine)
24991-53-5
MFCD00132004
Granular Solid
White
M.W. 1,000
H2NCH2CH2(OCH2CH2)nNH2
PEG-diamine
Air Sensitive
250mg
Chemical Identifiers
24991-53-5
H2NCH2CH2(OCH2CH2)nNH2
MFCD00132004
PEG-diamine
SDS Documents