Learn More
Polymixin B sulfate, Cell Culture Reagent, Thermo Scientific Chemicals
Polymixin B sulfate, CAS # 1405-20-5, is a short polypeptide with antibacterial activity against gram-negative bacteria.
$173.90 - $618.66
Chemical Identifiers
CAS | 1405-20-5 |
---|---|
Molecular Formula | C48H84N16O17S |
Molecular Weight (g/mol) | 1189.36 |
MDL Number | MFCD00131991 |
InChI Key | HNDFYNOVSOOGDU-UHFFFAOYNA-N |
PubChem CID | 133109994 |
SMILES | OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6176303
|
Thermo Scientific Chemicals
J6176303 |
1 g |
Each for $173.90
|
|
|||||
AAJ6176306
|
Thermo Scientific Chemicals
J6176306 |
5 g |
Each for $618.66
|
|
|||||
Description
In cell culture, polymyxin B sulfate is used as an antibacterial agent to prevent contamination. Polymyxin B sulfate is recommended for use in cell culture applications at 50 mg/L. It is used as a lipid A binding agent that induces pore formation in cell membranes and as immobilized (substrate bound) agent for removal of endotoxins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1405-20-5 | |
1189.36 | |
HNDFYNOVSOOGDU-UHFFFAOYNA-N | |
OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O |
C48H84N16O17S | |
MFCD00131991 | |
133109994 |
Specifications
Polymixin B Sulfate | |
1405-20-5 | |
MFCD00131991 | |
Soluble in water | |
OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O | |
1189.36 | |
1385.63 | |
Polymixin B sulfate |
Powder | |
C48H84N16O17S | |
Glass Bottle | |
HNDFYNOVSOOGDU-UHFFFAOYNA-N | |
N-[(2S)-4-amino-1-[[(2S,3R)-1-[[(2S)-4-amino-1-oxo-1-[[(3S,6S,18R)-6,9,18-tris(2-aminoethyl)-15-benzyl-3-[(1R)-1-hydroxyethyl]-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclotricos-21-yl]amino]butan-2-yl]amino]-3-hydroxy-1-ox | |
133109994 | |
1 g |
Safety and Handling
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302-H336
EINECSNumber : 215-774-7
RTECSNumber : TR150000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only