missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polymethylhydrosiloxane, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 175091000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Colorless | |
Viscous Liquid | |
(CH4OSi)n | |
MFCD00084478 | |
polymethyl hydrosiloxane, 2-methyl-1,2-oxasilinanato, ch3 3sio ch3 hsio nsi ch3 3 | |
100 g | |
C[SiH](-*)O-* | |
60.13 | |
121°C | |
Glass bottle | |
1 |
Polymethylhydrosiloxane | |
9004-73-3 | |
(CH3)3SiO[(CH3)HSiO]nSi(CH3)3 | |
1.0000g/mL | |
Solubility in water: practically insoluble. Other solubilities: may react, soluble in most common organic solvents, (should not be used in halocarbon solvent because, of the possibility of hydride-halide interchange, on catalysis or with heating) | |
TXYXVOOPRILIHR-UHFFFAOYSA-N | |
2-methyl-1,2$l^{3}-oxasilinane | |
12487658 | |
Authentic | |
1.3970 to 1.4010 | |
15-30 mm2/s (25°C) |
Chemical Identifiers
9004-73-3 | |
60.13 | |
TXYXVOOPRILIHR-UHFFFAOYSA-N | |
12487658 | |
C[SiH](-*)O-* |
(CH4OSi)n | |
MFCD00084478 | |
polymethyl hydrosiloxane, 2-methyl-1,2-oxasilinanato, ch3 3sio ch3 hsio nsi ch3 3 | |
2-methyl-1,2$l^{3}-oxasilinane |
RUO – Research Use Only