Please login to your online account to display your discounted pricing
Thermo Scientific™ Polymer Tubing Cutter
Produce flat, 90°, burr-free ends of polymer tubing for HPLC applications using Thermo Scientific™ Polymer Tubing Cutter.
Manufacturer: thermo scientific™  A327
 View more versions of this product
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Catalog No. 03-052-399
  • / Each
Description & Specifications
Specifications
Description 90° Burr-Free End
Quantity 1 Each
For Use With Tubing
Type Tubing Cutter
Produce flat, 90°, burr-free ends of polymer tubing for HPLC applications using the Thermo Scientific™ Polymer Tubing Cutter. It is compatible with rigid polymeric tubing and comes with guide holes for 1/16" and 1/8" tubing.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.