Please login to your online account to display your discounted pricing

Polymedco™ Sediplast™ Autozero ESR System


Closed system with unique vented cap: precisely calibrated hole stops the column of blood at true “zero”

Manufacturer: polymedco inc  S1090*

 View more versions of this product

Catalog No. 02-681-14

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Includes Thin diaphragm prefilled vial containing 0.2mL of 3.8% Sodium Citrate
For Use With (Application) Used for hematology applications
Product Type Autozero ESR system
Sample Type Whole EDTA Blood