Polymedco™ Sediplast™ Autozero ESR System
Closed system with unique vented cap: precisely calibrated hole stops the column of blood at true “zero”
Manufacturer:  Polymedco Inc S1030
 View more versions of this product
Catalog No. 02-670-28
Specifications Description & Specifications
Specifications
SEDIPLAST Autozero ESR System
Whole EDTA Blood
Used for hematology applications
SediPlast leveling plate