Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyimide resin

Soluble thermoplastic polyimide for high temperature adhesives and composites

$51.68 - $515.31

Chemical Identifiers

Chemical Name or Material Polyimide resin
CAS 62929-02-6
MDL Number MFCD01868102
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4365609

  SDS
alfa aesar™
4365609
10g Each for $51.68

AA4365630

  SDS
alfa aesar™
4365630
250g Each for $515.31
Description & Specifications

Specifications

Health Hazard 1 Warning
TSCA TSCA
Quantity 10g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Color Yellow
Density 1.2g/mL
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Polyimide resin
Flash Point >93°C (199°F)
MDL Number MFCD01868102
CAS 62929-02-6
Physical Form Powder
Melting Point >300°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.