missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyimide resin

Soluble thermoplastic polyimide for high temperature adhesives and composites

Manufacturer:  Alfa Aesar™ 4365630

 View more versions of this product

Catalog No. AA4365630



Specifications

Specifications

Polyimide resin
1.2g/mL
MFCD01868102
CDTDIQLZIBORMV-UHFFFAOYSA-N
1-(4-aminophenyl)-1,3,3-trimethyl-2H-inden-5-amine;5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione
6454485
Yellow
250g
62929-02-6
C35H28N2O7
1,3-isobenzofurandione, 5,5'-carbonylbis-, polymer with 1 or 3-4-aminophenyl-2,3-dihydro-1,3,3 or 1,1,3-trimethyl-1h-inden-5-amine
CC1(CC(C2=C1C=C(C=C2)N)(C)C3=CC=C(C=C3)N)C.C1=CC2=C(C=C1C(=O)C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O
588.616
Powder
>300°C
>93°C (199°F)

Chemical Identifiers

62929-02-6
588.616
CDTDIQLZIBORMV-UHFFFAOYSA-N
6454485
CC1(CC(C2=C1C=C(C=C2)N)(C)C3=CC=C(C=C3)N)C.C1=CC2=C(C=C1C(=O)C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O
C35H28N2O7
MFCD01868102
1,3-isobenzofurandione, 5,5'-carbonylbis-, polymer with 1 or 3-4-aminophenyl-2,3-dihydro-1,3,3 or 1,1,3-trimethyl-1h-inden-5-amine
1-(4-aminophenyl)-1,3,3-trimethyl-2H-inden-5-amine;5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents