Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyimide film, 0.025mm thick x 20mm width 33m

Chemical Identifiers

Chemical Name or Material Polyimide
CAS 62929-02-6
Name Note 0.025mm thick x 20mm width
MDL Number MFCD00677744
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41898UL

 
alfa aesar™
41898UL
33m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 33m
CAS 62929-02-6
Name Note 0.025mm thick x 20mm width
Physical Form Film
Chemical Name or Material Polyimide
MDL Number MFCD00677744