Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethyleneimine on silica beads, anion exchange resin, benzylated, 40-200 mesh

Solid phase organic synthesis, affinity chromatography, catalyst support

$38.12 - $326.97

Chemical Identifiers

Chemical Name or Material Polyethyleneimine on silica beads anion exchange resin
CAS 9002-98-6
Name Note Benzylated
MDL Number MFCD00084427
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43305AB

  SDS
alfa aesar™
43305AB
10mL Each for $38.12

AA43305AK

  SDS
alfa aesar™
43305AK
250mL Each for $326.97
Description & Specifications

Specifications

MDL Number MFCD00084427
Sensitivity Hygroscopic
Quantity 10mL
Chemical Name or Material Polyethyleneimine on silica beads anion exchange resin
Mesh Size 40 to 200 Mesh
Name Note Benzylated
Physical Form Powder
CAS 9002-98-6