missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethyleneimine on silica beads, anion exchange resin, benzylated, 20-40 mesh

Solid phase organic synthesis, affinity chromatography, catalyst support

$46.93 - $356.80

Chemical Identifiers

CAS 9002-98-6
Molecular Formula C2H5N
Molecular Weight (g/mol) 43.069
MDL Number MFCD00084427
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
Synonym azacyclopropane, aziran, dihydroazirene, dimethyleneimine, ethylene imine, ethyleneimine, ethylenimine, everamine, polyethyleneimine, polymin
PubChem CID 9033
ChEBI CHEBI:30969
IUPAC Name aziridine
SMILES C1CN1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA43304AB
View Documents
Alfa Aesar™
43304AB
10mL Each for $46.93
 
AA43304AK
View Documents
Alfa Aesar™
43304AK
250mL Each for $356.80
 
Specifications

Chemical Identifiers

9002-98-6
43.069
NOWKCMXCCJGMRR-UHFFFAOYSA-N
9033
aziridine
C2H5N
MFCD00084427
azacyclopropane, aziran, dihydroazirene, dimethyleneimine, ethylene imine, ethyleneimine, ethylenimine, everamine, polyethyleneimine, polymin
CHEBI:30969
C1CN1

Specifications

9002-98-6
MFCD00084427
NOWKCMXCCJGMRR-UHFFFAOYSA-N
aziridine
9033
Beads
Hygroscopic
Polyethyleneimine on silica beads anion exchange resin, Benzylated
C2H5N
azacyclopropane, aziran, dihydroazirene, dimethyleneimine, ethylene imine, ethyleneimine, ethylenimine, everamine, polyethyleneimine, polymin
C1CN1
43.069
CHEBI:30969
20 to 40 Mesh
10mL
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures