missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethyleneimine on silica beads, anion exchange resin, benzylated, 20-40 mesh

Solid phase organic synthesis, affinity chromatography, catalyst support

Manufacturer:  Alfa Aesar™ 43304AB

 View more versions of this product

Catalog No. AA43304AB



Specifications

Specifications

Polyethyleneimineonsilicabeadsanionexchangeresin
9002-98-6
MFCD00084427
NOWKCMXCCJGMRR-UHFFFAOYSA-N
aziridine
9033
Beads
Hygroscopic
Benzylated
C2H5N
azacyclopropane, aziran, dihydroazirene, dimethyleneimine, ethylene imine, ethyleneimine, ethylenimine, everamine, polyethyleneimine, polymin
C1CN1
43.069
CHEBI:30969
20 to 40 Mesh
10mL

Chemical Identifiers

9002-98-6
43.069
NOWKCMXCCJGMRR-UHFFFAOYSA-N
9033
aziridine
C2H5N
MFCD00084427
azacyclopropane, aziran, dihydroazirene, dimethyleneimine, ethylene imine, ethyleneimine, ethylenimine, everamine, polyethyleneimine, polymin
CHEBI:30969
C1CN1
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents