Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethyleneimine on silica beads, anion exchange resin, 40-200 mesh

For the separation of nucleotide mixtures

$40.47 - $363.27

Chemical Identifiers

Chemical Name or Material Polyethyleneimine on silica beads anion exchange resin
CAS 9002-98-6
MDL Number MFCD00084427
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43303AB

  SDS
alfa aesar™
43303AB
10mL Each for $40.47

AA43303AK

  SDS
alfa aesar™
43303AK
250mL Each for $363.27

AA43303AD

  SDS
alfa aesar™
43303AD
50mL Each for $104.36
Description & Specifications

Specifications

Chemical Name or Material Polyethyleneimine on silica beads anion exchange resin
TSCA TSCA
Physical Form Powder
MDL Number MFCD00084427
CAS 9002-98-6
Mesh Size 40 to 200 Mesh
Sensitivity Hygroscopic
Quantity 10mL