Alfa Aesar™ Polyethyleneimine on silica beads, anion exchange resin, 20-40 mesh
For the separation of nucleotide mixtures
Manufacturer:  Alfa Aesar™ 43302AK
 View more versions of this product
Catalog No. AA43302AK
Specifications
Specifications
Polyethyleneimine on silica beads anion exchange resin
C2H5N
azacyclopropane
C1CN1
43.069
CHEBI:30969
20 to 40 Mesh
250mL
9002-98-6
MFCD00084427
NOWKCMXCCJGMRR-UHFFFAOYSA-N
aziridine
9033
Beads
Hygroscopic
Chemical Identifiers
9002-98-6
43.069
NOWKCMXCCJGMRR-UHFFFAOYSA-N
9033
aziridine
C2H5N
MFCD00084427
azacyclopropane
CHEBI:30969
C1CN1
Safety and Handling
Safety and Handling
TSCA : TSCA
Documents