missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethyleneimine, branched, M.W. 50,000 - 100,000, 30% w/w aq. soln.

Manufacturer:  Alfa Aesar™ 4502414

 View more versions of this product

Catalog No. AA4502414



Specifications

Specifications

Polyethyleneimine
9002-98-6
MFCD00803910
NOWKCMXCCJGMRR-UHFFFAOYSA-N
aziridine
9033
Liquid
Colorless to Yellow
25g
Branched, M.W. 50,000 to 100,000
C2H5N
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
C1CN1
43.069
CHEBI:30969
Odorless
30% w/w aq. soln.

Chemical Identifiers

9002-98-6
43.069
NOWKCMXCCJGMRR-UHFFFAOYSA-N
9033
aziridine
C2H5N
MFCD00803910
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
CHEBI:30969
C1CN1
Safety and Handling

Safety and Handling

RTECSNumber : TQ7915000

TSCA : TSCA

SDS Documents