missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethyleneimine, branched, M.W. 1,200, 99%

Manufacturer:  Alfa Aesar™ 4559414

 View more versions of this product

Catalog No. AA4559414



Specifications

Specifications

Polyethyleneimine
9002-98-6
MFCD00803910
NOWKCMXCCJGMRR-UHFFFAOYSA-N
aziridine
9033
Liquid
Amine-like
25g
Branched, M.W. 1,200
C2H5N
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
C1CN1
43.069
CHEBI:30969
99%
Yellow

Chemical Identifiers

9002-98-6
43.069
NOWKCMXCCJGMRR-UHFFFAOYSA-N
9033
aziridine
C2H5N
MFCD00803910
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
CHEBI:30969
C1CN1
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents